Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-4 Antibody (MM0300-1D12), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212323F
Description
Galectin-4 Monoclonal specifically detects Galectin-4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Galectin-4 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
LGALS4 | |
Human recombinant Galectin-4 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0300-1D12 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Antigen NY-CO-27, gal-4, GAL4, galectin 4, galectin-4, L-36 lactose-binding protein, L36LBP, Lactose-binding lectin 4, lectin, galactoside-binding, soluble, 4 | |
Mouse | |
Protein G purified | |
Cancer | |
3960 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction