Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-4 Antibody (SR1598), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321804100UL
Description
Galectin-4 Monoclonal antibody specifically detects Galectin-4 in Human, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Galectin-4 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
SR1598 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
Antigen NY-CO-27, gal-4, GAL4, galectin 4, galectin-4, L-36 lactose-binding protein, L36LBP, Lactose-binding lectin 4, lectin, galactoside-binding, soluble, 4 | |
A synthesized peptide derived from human Galectin-4 (Uniprot #: P56470) | |
100 μg | |
Cancer | |
3960 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction