Learn More
Invitrogen™ Galectin 4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595353
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: SW620 whole cell, COLO320 whole cell. ICC/IF: A431 cell. Flow: THP-1 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The expression of this gene is restricted to small intestine, colon, and rectum, and it is underexpressed in colorectal cancer. Galectin 4 binds lactose and a related range of sugars. May be involved in the assembly of adherens junctions.
Specifications
Galectin 4 | |
Polyclonal | |
Unconjugated | |
Lgals4 | |
Antigen NY-CO-27; GAL4; gal-4; galectin 4; galectin-4; L-36; L-36 lactose-binding protein; L36LBI; L36LBP; lactose-binding lectin 4; Lectin galactose binding soluble 4 (Galectin-4); lectin, galactose binding, soluble 4; Lectin, galactose binding, soluble 4 (Galectin-4); lectin, galactoside binding soluble 4; lectin, galactoside-binding, soluble, 4; lectin, galactoside-binding, soluble, 4 (galectin 4); Lgals4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3960 | |
-20°C | |
Lyophilized |
Flow Cytometry, Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
P56470 | |
Lgals4 | |
A synthetic peptide corresponding to a sequence at the C-terminus of human GAL4 (283-320aa DRFKVYANGQHLFDFAHRLSAFQRVDTLEIQGDVTLSY). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.