Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Galectin 5 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. FEPA5145325
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145325 200 μL
FEPA5145326 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145325 Supplier Invitrogen™ Supplier No. PA5145325
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Galectin 5 is a member of the galectin family of proteins, which are characterized by their ability to bind to specific carbohydrate structures on cell surfaces. Galectin 5 is primarily expressed in the cytoplasm of cells and has been shown to play a role in regulating cell growth, differentiation, and apoptosis. It has also been implicated in various physiological processes, including inflammation, immune response, and cancer progression. Galectin 5 has been found to interact with a variety of proteins and signaling pathways, suggesting that it may have multiple functions within cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin 5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene Lgals5
Gene Accession No. P47967
Gene Alias Lgals5
Gene Symbols Lgals5
Host Species Rabbit
Immunogen Recombinant Rat GAL5 protein, Met1-Thr145 (Accession #P47967).
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25475
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.