Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-7 Antibody (950723) [Allophycocyanin], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB13392A
Description
Galectin-7 Monoclonal antibody specifically detects Galectin-7 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOFSpecifications
Galectin-7 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
950723 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready | |
Gal-7, GAL7, galectin 7, galectin-7, HKL-14, lectin, galactoside-binding, soluble, 7, LGALS7A, p53-induced gene 1 protein, PI7, PIG1, TP53I1 | |
E. coli-derived recombinant human Galectin-7, Ser2-Phe136, Accession # NP_002298 | |
0.1 mL | |
Apoptosis, Cancer, Tumor Suppressors | |
3963 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction