Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-7 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100380AF350
Description
Galectin-7 Polyclonal antibody specifically detects Galectin-7 in Mouse samples. It is validated for Western BlotSpecifications
Galectin-7 | |
Polyclonal | |
Western Blot | |
Gal-7, GAL7, galectin 7, galectin-7, HKL-14, lectin, galactoside-binding, soluble, 7, LGALS7A, p53-induced gene 1 protein, PI7, PIG1, TP53I1 | |
The immunogen recognized by this antibody maps to a region between residue 100 and the C-terminus (residue 135) of human Galectin-7 using the numbering given in entry NP_002298.1 (GeneID 3963). | |
0.1 mL | |
Apoptosis, Cancer, Tumor Suppressors | |
3963 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction