Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Galectin 7 Monoclonal Antibody (C2), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA543813
This item is not returnable.
View return policy
Description
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. Differential and in situ hybridization studies indicate that this lectin is specifically expressed in keratinocytes and found mainly in stratified squamous epithelium. A duplicate copy of this gene (GeneID:653499) is found adjacent to, but on the opposite strand on chromosome 19.
Specifications
Galectin 7 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P47929, P97590 | |
LGALS7, LGALS7B | |
Met1-Phe136 (Accession P47929), with N-terminal His Tag. | |
20 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 κ |
Immunohistochemistry (Paraffin), Western Blot | |
C2 | |
Unconjugated | |
LGALS7 | |
GAL7; Gal-7; galectin 7; galectin 7B; galectin-7; galectin-7B; HKL-14; keratinocyte lectin 14; lectin galactoside-binding soluble 7; lectin, galactose binding, soluble 7; lectin, galactoside binding soluble 7; lectin, galactoside binding soluble 7B; lectin, galactoside-binding, soluble, 7; lectin, galactoside-binding, soluble, 7B; LGALS7; LGALS7A; LGALS7B; p53-induced gene 1 protein; p53-induced protein 1; PI7; Pig1 | |
Mouse | |
Protein A/G | |
RUO | |
29518, 3963, 653499 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction