Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Galectin 7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547124
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547124 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547124 Supplier Invitrogen™ Supplier No. PA547124
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In sandwich ELISAs, approximately 50% cross-reactivity with recombinant mouse (rm) Galectin-7 is observed and less than 0.5% cross-reactivity with recombinant human (rh) Galectin-1, rmGalectin-3, rhGalectin-4, and rhGalectin-8 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. Differential and in situ hybridization studies indicate that this lectin is specifically expressed in keratinocytes and found mainly in stratified squamous epithelium. A duplicate copy of this gene (GeneID:653499) is found adjacent to, but on the opposite strand on chromosome 19.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin 7
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene LGALS7
Gene Accession No. P47929
Gene Alias GAL7; Gal-7; galectin 7; galectin 7B; galectin-7; galectin-7B; HKL-14; keratinocyte lectin 14; lectin galactoside-binding soluble 7; lectin, galactose binding, soluble 7; lectin, galactoside binding soluble 7; lectin, galactoside binding soluble 7B; lectin, galactoside-binding, soluble, 7; lectin, galactoside-binding, soluble, 7B; LGALS7; LGALS7A; LGALS7B; p53-induced gene 1 protein; p53-induced protein 1; PI7; Pig1
Gene Symbols LGALS7, LGALS7B
Host Species Goat
Immunogen E. coli-derived recombinant human Galectin-7 Ser2-Phe136.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3963, 653499
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.