Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Galectin 7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581953
Description
Immunogen sequence: PHKSSLPEGI RPGTVLRIRG LVPPNASRFH VNLLCGEEQG SDAALHFNPR LDTSEVVFNS KEQGSWGREE RGPGVPFQRG QPFEVLIIAS DDGFKAVVGD AQYHHFRHRL PLARVRLVEV GGDVQLDSV.
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. Differential and in situ hybridization studies indicate that this lectin is specifically expressed in keratinocytes and found mainly in stratified squamous epithelium. A duplicate copy of this gene (GeneID:653499) is found adjacent to, but on the opposite strand on chromosome 19.
Specifications
Galectin 7 | |
Polyclonal | |
Unconjugated | |
LGALS7 | |
GAL7; Gal-7; galectin 7; galectin 7B; galectin-7; galectin-7B; HKL-14; keratinocyte lectin 14; lectin galactoside-binding soluble 7; lectin, galactose binding, soluble 7; lectin, galactoside binding soluble 7; lectin, galactoside binding soluble 7B; lectin, galactoside-binding, soluble, 7; lectin, galactoside-binding, soluble, 7B; LGALS7; LGALS7A; LGALS7B; p53-induced gene 1 protein; p53-induced protein 1; PI7; Pig1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3963, 653499 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P47929 | |
LGALS7, LGALS7B | |
Recombinant protein corresponding to Human LGALS7. Recombinant protein control fragment (Product #RP-94760). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction