Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Galectin 7 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. PIPA5121968
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121968 100 μL
PIPA5121969 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121968 Supplier Invitrogen™ Supplier No. PA5121968
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. Differential and in situ hybridization studies indicate that this lectin is specifically expressed in keratinocytes and found mainly in stratified squamous epithelium. A duplicate copy of this gene (GeneID:653499) is found adjacent to, but on the opposite strand on chromosome 19.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin 7
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene LGALS7
Gene Accession No. O54974, P47929, P97590
Gene Alias GAL7; Gal-7; galectin 7; galectin 7B; galectin-7; galectin-7B; HKL-14; keratinocyte lectin 14; lectin galactoside-binding soluble 7; lectin, galactose binding, soluble 7; lectin, galactoside binding soluble 7; lectin, galactoside binding soluble 7B; lectin, galactoside-binding, soluble, 7; lectin, galactoside-binding, soluble, 7B; LGALS7; LGALS7A; LGALS7B; p53-induced gene 1 protein; p53-induced protein 1; PI7; Pig1
Gene Symbols LGALS7, LGALS7B
Host Species Rabbit
Immunogen Met1-Phe136 (Accession P47929), with N-terminal His Tag.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16858, 29518, 3963, 653499
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.