Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 167301AP150UL

Description
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. Differential and in situ hybridization studies indicate that this lectin is specifically expressed in keratinocytes and found mainly in stratified squamous epithelium. A duplicate copy of this gene (GeneID:653499) is found adjacent to, but on the opposite strand on chromosome 19.Specifications
Galectin-7 | |
Polyclonal | |
Unconjugated | |
LGALS7B | |
Gal 7, GAL7, Galectin 7, Galectin-7, HKL 14, LGALS7, LGALS7B, p53 induced gene 1 protein, PI7, PIG1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
653499 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P47929 | |
LGALS7B | |
Galectin-7 Fusion Protein Ag10188 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction