Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-8 Antibody (106.1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311211
Description
Galectin-8 Monoclonal specifically detects Galectin-8 in Human, Rat samples. It is validated for Flow Cytometry, Functional, Immunoassay.Specifications
Galectin-8 | |
Monoclonal | |
0.1 mg/ml | |
Flow Cytometry, Functional, Immunoassay | |
Gal-8, galectin 8, galectin-8, galectin-8g, lectin, galactoside-binding, soluble, 8, PCTA1, PCTA-1Po66 carbohydrate-binding protein, Po66 carbohydrate binding protein, Po66-CBP, Prostate carcinoma tumor antigen 1 | |
Recombinant rat Galectin-8 | |
50 μg | |
Primary | |
Human, Rat | |
Purified |
Flow Cytometry, Functional Assay, Immunoassay | |
106.1 | |
Unconjugated | |
0.2 um filtered solution in PBS, 0.1% BSA | |
Mouse | |
Protein G purified | |
RUO | |
3964 | |
Store at 4°C. Do not freeze. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction