Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Galectin 8 Monoclonal Antibody (106.1)
GREENER_CHOICE

Catalog No. PIMA533349
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533349 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533349 Supplier Invitrogen™ Supplier No. MA533349
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Product is supplied at 1 mL (100 μL/mL).

This gene encodes a member of the galectin family. Galectins are beta-galactoside-binding animal lectins with conserved carbohydrate recognition domains. The galectins have been implicated in many essential functions including development, differentiation, cell-cell adhesion, cell-matrix interaction, growth regulation, apoptosis, and RNA splicing. This gene is widely expressed in tumoral tissues and seems to be involved in integrin-like cell interactions. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin 8
Applications ELISA, Flow Cytometry, Functional Assay
Classification Monoclonal
Clone 106.1
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and no preservative
Gene Lgals8
Gene Accession No. Q62665
Gene Alias 1200015E08Rik; 30 kDa S-type lectin; AI326142; D13Ertd524e; Gal-8; galectin 8; galectin-8; galectin-8g; galectin-8L; lectin, galactose binding, soluble 5; lectin, galactose binding, soluble 8; lectin, galactoside binding soluble 8; lectin, galactoside-binding, soluble, 8; LGALS8; Lgals-8; PCTA1; PCTA-1; Po66 carbohydrate binding protein; po66 carbohydrate-binding protein; Po66-CBP; prostate carcinoma tumor antigen 1; RL-30; RP11-385F5.1
Gene Symbols Lgals8
Host Species Mouse
Immunogen recombinant rat Galectin-8.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116641
Target Species Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.