Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Galectin 8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528506
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528506 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528506 Supplier Invitrogen™ Supplier No. PA528506
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T transfected recombinant LGALS8 protein. Predicted reactivity: Rat (80%), Dog (86%), Pig (83%), Rhesus Monkey (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the galectin family. Galectins are beta-galactoside-binding animal lectins with conserved carbohydrate recognition domains. The galectins have been implicated in many essential functions including development, differentiation, cell-cell adhesion, cell-matrix interaction, growth regulation, apoptosis, and RNA splicing. This gene is widely expressed in tumoral tissues and seems to be involved in integrin-like cell interactions. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin 8
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Lgals8
Gene Accession No. O00214
Gene Alias 1200015E08Rik; 30 kDa S-type lectin; AI326142; D13Ertd524e; Gal-8; galectin 8; galectin-8; galectin-8g; galectin-8L; lectin, galactose binding, soluble 5; lectin, galactose binding, soluble 8; lectin, galactoside binding soluble 8; lectin, galactoside-binding, soluble, 8; LGALS8; Lgals-8; PCTA1; PCTA-1; Po66 carbohydrate binding protein; po66 carbohydrate-binding protein; Po66-CBP; prostate carcinoma tumor antigen 1; RL-30; RP11-385F5.1
Gene Symbols Lgals8
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Galectin 8. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3964
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.