Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Galectin 8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595297
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595297 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595297 Supplier Invitrogen™ Supplier No. PA595297
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human COLO-320 whole cell, rat testicular tissue, mouse Neuro-2A whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

This gene encodes a member of the galectin family. Galectins are beta-galactoside-binding animal lectins with conserved carbohydrate recognition domains. The galectins have been implicated in many essential functions including development, differentiation, cell-cell adhesion, cell-matrix interaction, growth regulation, apoptosis, and RNA splicing. This gene is widely expressed in tumoral tissues and seems to be involved in integrin-like cell interactions. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin 8
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene Lgals8
Gene Accession No. O00214, Q62665, Q9JL15
Gene Alias 1200015E08Rik; 30 kDa S-type lectin; AI326142; D13Ertd524e; Gal-8; galectin 8; galectin-8; galectin-8g; galectin-8L; lectin, galactose binding, soluble 5; lectin, galactose binding, soluble 8; lectin, galactoside binding soluble 8; lectin, galactoside-binding, soluble, 8; LGALS8; Lgals-8; PCTA1; PCTA-1; Po66 carbohydrate binding protein; po66 carbohydrate-binding protein; Po66-CBP; prostate carcinoma tumor antigen 1; RL-30; RP11-385F5.1
Gene Symbols Lgals8
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human Galectin 8 (286-317aa HSLEYKHRFKELSSIDTLEINGDIHLLEVRSW).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116641, 3964, 56048
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.