Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-9 Antibody (766428) [Alexa Fluor™ 532], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB3535X
Description
Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Mouse samples. It is validated for Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
Galectin-9 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF | |
766428 | |
Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
ecalectin, Gal-9, galectin 9, galectin-9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein | |
E. coli-derived recombinant mouse Galectin-9 (short isoform), Ala2-Thr322, Accession # O08573-2 | |
0.1 mL | |
Myeloid derived Suppressor Cell, Signal Transduction | |
3965 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction