Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-9 Antibody (9M1-3), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP318530MFV610
Description
Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Human, Primate samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
Galectin-9 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Primate | |
Purified |
Flow Cytometry | |
9M1-3 | |
Flow Cytometry, Flow (Intracellular) | |
ecalectin, Gal-9, galectin 9, galectin-9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein | |
Recombinant M-type splicing variant of human Galectin-9 | |
0.1 mL | |
Myeloid derived Suppressor Cell, Signal Transduction | |
3965 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction