Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-9 Antibody (OTI1D12), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271132G
Description
Galectin-9 Monoclonal specifically detects Galectin-9 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Galectin-9 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LGALS9 | |
Full length recombinant protein of human Galectin-9 (NP_002299) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI1D12 | |
Western Blot, Immunohistochemistry | |
ecalectin, Gal-9, galectin 9, galectin-9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein | |
Mouse | |
Affinity Purified | |
Myeloid derived Suppressor Cell, Signal Transduction | |
3965.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction