Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Galectin-9 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems IC3535V100UG
This item is not returnable.
View return policy
Description
Galectin-9 Monoclonal specifically detects Galectin-9 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Galectin-9 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
O08573-2 | |
LGALS9 | |
E. coli-derived recombinant mouse Galectin-9 (short isoform) Ala2-Thr322 Accession # O08573-2 | |
RUO | |
3965 | |
Mouse | |
IgG2b |
Flow Cytometry | |
766428 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
ecalectin, gal-9, GAL9, galectin 9, galectin-9, Galectin9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein | |
Rat | |
100 μg | |
Primary | |
Detects mouse Galectin-9 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Galectin-9 or recombinant mouse Galectin-4 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction