Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin-9 Rat anti-Mouse, BV421, Clone: RG9-35, BD Horizon™

Catalog No. BDB566028 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566028 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566028 Supplier BD Biosciences Supplier No. 566028
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Mouse samples. It is validated for Flow cytometry (Intracellular Staining)) .
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin-9
Applications Flow Cytometry, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone RG9-35
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias Galectin9; Gal-9; Gal9; Lgals9; LEG9
Host Species Rat
Immunogen Mouse Galectin-9 Recombinant Protein
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Reseach Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.