Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Galeterone
GREENER_CHOICESDP

Catalog No. 610850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-085-0 50 mg
61-081-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 61-085-0 Supplier Tocris Bioscience™ Supplier No. 6108/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

CYP17 inhibitor; also androgen receptor antagonist

Galeterone is a CYP17 inhibitor (IC50 = 300 nM) and androgen receptor antagonist (IC50 = 384 nM in PC3-AR cells). Inhibits DHT-stimulated prostate cancer cell proliferation in vitro. Exhibits antitumor activity in SCID mice bearing prostate cancer xenografts.

Specifications

Chemical Name or Material Galeterone
CAS 851983-85-2
Synonym (3β)-17-(1H-Benzimidazol-1-yl)androsta-5,16-dien-3-ol
Quantity 50 mg
Solubility Soluble to 50mM in DMSO and to 50mM in ethanol
Molecular Formula C26H32N2O
Formula Weight 388.55
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.