Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GALK1 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299783AF594
Description
GALK1 Polyclonal specifically detects GALK1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
GALK1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
2584 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
Alexa Fluor 594 | |
EC 2.7.1.6, galactokinase, galactokinase 1, Galactose kinase, GALK, GK1 | |
Produced in rabbits immunized with purified, recombinant GALK1 (P51570; Met1-Leu392). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction