Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GALK1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586708
Description
Purity is > 95% by SDS-PAGE.
Galactokinase is a major enzyme for the metabolism of galactose and its deficiency causes congenital cataracts in the adult population. GALK1 sequence shares the greatest level of conservation, 44. 5% identity with that from E. coli and 34. 6% amino acid identity with the product of the human GALK2 gene.
Specifications
GALK1 | |
Polyclonal | |
Unconjugated | |
GALK1 | |
AA409894; epididymis secretory protein Li 19; galactokinase; galactokinase 1; galactose kinase; GALK; Galk1; Gk; GK1; Glk; Glk1; HEL-S-19 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14635, 2584, 287835 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P51570, Q9R0N0 | |
GALK1 | |
Synthetic peptide, corresponding to amino acids 4-49 of Human GALK1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction