Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GALK1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304421100UL
Description
GALK1 Polyclonal specifically detects GALK1 in Human, Mouse samples. It is validated for Western Blot.Specifications
GALK1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
2584 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
EC 2.7.1.6, galactokinase, galactokinase 1, Galactose kinase, GALK, GK1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human GALK1 (NP_000145.1). | |
100 μL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction