Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GALK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583668
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583668 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583668 Supplier Invitrogen™ Supplier No. PA583668
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVDICRKFGA QGSRLTGAGW GGCTVSMVPA DKLPSFLANV HKAYYQRSDG SLAPEKQSLF ATKPGGGALV LLE Highest antigen sequence indentity to the following orthologs: Rat - 81%, Mouse - 79%.

This gene encodes a highly efficient N-acetylgalactosamine kinase, which has galactokinase activity when galactose is present at high concentrations. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALK2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GALK2
Gene Accession No. Q01415
Gene Alias 2810017M24Rik; AI327030; Galactokinase 2; GALK2; GalNAc kinase; Gk2; N-acetylgalactosamine kinase
Gene Symbols GALK2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GALK2. Recombinant protein control fragment (Product #RP-98118).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2585
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.