Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Gallein
GREENER_CHOICESDP

Catalog No. 309050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-905-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 30-905-0 Supplier Tocris Bioscience™ Supplier No. 3090/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibitor of βγ signaling

Inhibitor of G protein βγ subunit-dependent signaling. Blocks PI 3-kinase and Rac1 activation in HL60 cells and chemotaxis in HL60 differentiated cells.

Chemical Identifiers

CAS 2103-64-2
Molecular Formula C20H12O7

Specifications

Quantity 50 mg
Formula Weight 364.31
Purity >98%
Molecular Weight (g/mol) 364.309
Chemical Name or Material Gallein
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.