Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gallium(III) bromide, ultra dry, 99.998% (metals basis)
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Gallium bromide is used as a catalyst in the alkylation of aromatic compounds by alkyl bromides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon

Specifications
Specifications
Boiling Point | 279°C |
Melting Point | 122°C |
Color | White |
Physical Form | Crystalline |
Quantity | 5 g |
UN Number | UN3260 |
Solubility Information | Soluble in water. |
Formula Weight | 309.45 |
Percent Purity | 99.998% |
Odor | Odorless |
Show More |
Safety and Handling
Hazard Category | H314 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: III |
EINECSNumber | 236-609-5 |
TSCA | Yes |
storageNote1 |
|
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction