Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallium(III) chloride, 99.99+%, (trace metal basis), Anhydrous, Beads, -10 mesh

Catalog No. p-4440167
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 13450-90-3 | Cl3Ga | 176.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13450-90-3
Molecular Formula Cl3Ga
Molecular Weight (g/mol) 176.08
InChI Key UPWPDUACHOATKO-UHFFFAOYSA-K
Synonym gallium trichloride, gallium iii chloride, trichlorogallium, gallium chloride, gallium chloride gacl3, gallium 3+ chloride, gacl3, ccris 4492, gallium iii chloride, ultra dry, trichlorogalliue
PubChem CID 26010
IUPAC Name trichlorogallane
SMILES Cl[Ga](Cl)Cl

Specifications

Boiling Point 201.0°C
Melting Point 78.0°C
Quantity 5 g
Assay Percent Range 99.99+%
Linear Formula GaCl3
Merck Index 15,4378
Solubility Information Solubility in water: reacts
Formula Weight 176.08
Percent Purity ≥99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Gallium(III) Chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Store in a d
EINECSNumber 236-610-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.