Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallium(III) fluoride, anhydrous, 99.85% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7049965
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g

Gallium(III) fluoride reacts with mineral acids to give hydrofluoric acid. It is used in the preparation and study of optical fluorogallate glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-51-9
MDL Number MFCD00011021

Specifications

Boiling Point ∼1000°C
Melting Point 800°C (sublimation)
Color White
Physical Form Powder
Quantity 5 g
UN Number UN3260
Merck Index 14,4354
Solubility Information Insoluble in water.
Formula Weight 126.72
Percent Purity 99.85%
Odor Odorless
Sensitivity Moisture sensitive
Density 4.47 g/mL
Assay (metals basis)
Chemical Name or Material Gallium(III) fluoride, Anhydrous
Show More Show Less
missing translation for 'healthHazard1' H314-H335
missing translation for 'healthHazard2' GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
missing translation for 'healthHazard3' P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 232-004-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.