Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gallium(III) iodide, ultra dry, 99.999% (metals basis)
Click to view available options
Quantity:
1 g
5 g
25 g
Description
It is useful for the preparation of compounds of gallium(I) and gallium(II) and is reported as useful in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon

Specifications
Specifications
Boiling Point | 345°C (sublimation) |
Melting Point | 212°C |
Color | Yellow to Green |
Physical Form | Beads |
Quantity | 25 g |
UN Number | UN3260 |
Solubility Information | Decomposes in water. |
Formula Weight | 450.43 |
Percent Purity | 99.999% |
Packaging | Ampule |
Show More |
Safety and Handling
Hazard Category | H314-H317-H334-H335 |
Hazard Statement | GHS H Statement H334-H314-H318-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. May cause an allergic skin reaction. |
Precautionary Statement | P260-P264b-P271-P272-P280-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. |
EINECSNumber | 236-611-6 |
TSCA | Yes |
storageNote1 |
|
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction