Learn More
Gallium(III) iodide, ultra dry, 99.999% (metals basis)
Description
Gallium(III) iodide is widely used as lewis acid, a precursor of organogallium reagents. It can also be involved in detection of solar neutrinos.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 345°C (sublimation) |
| Melting Point | 212°C |
| Color | Yellow to Green |
| Physical Form | Powder |
| Quantity | 5 g |
| Assay Percent Range | 99.999% (Metal basis) |
| Solubility Information | Soluble in aromatic solvents. |
| Formula Weight | 450.43 |
| Sensitivity | Light and moisture sensitive |
| Appearance | Yellow-green powder |
| Show More |
Safety and Handling
| Hazard Category | H314-H317-H334-H335 |
| Precautionary Statement | P260-P264b-P271-P272-P280-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c |
| DOTInformation | DOT Class: 8, Packing Group: II |
| EINECSNumber | 236-611-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.