Learn More
Gallium(III) isopropoxide, mixture of oligomers, 99%
Description
Gallium(III) isopropoxide can be used to accelerate the drying of paints or printing inks. It is also used as effective catalysts in the conversion of an aldehyde to an ester, ketones to secondary alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 72°C |
| Color | Yellow |
| Density | 1.35 g/mL |
| Boiling Point | 120°C (1 mm) |
| Flash Point | 68°C (154°F) |
| Linear Formula | Ga(OC3H7)3 |
| Quantity | 5 g |
| UN Number | UN1993 |
| Sensitivity | Moisture sensitive |
| Solubility Information | It reacts with water. |
| Show More |
Safety and Handling
| Hazard Category | H227-H315-H319 |
| Hazard Statement | GHS H Statement H226-H315-H319 Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.