Alfa Aesar™ Gallium(III) sulfate hydrate, Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1115209

Quantity 10g

 View more versions of this product

Catalog No. AA1115209

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13780-42-2
    Formula Weight 427.62 (Anhydrous)
    MDL Number MFCD00149725
    Chemical Name or Material Gallium(III) sulfate hydrate
    Grade Puratronic™

    Specifications

    Formula Weight 427.62 (Anhydrous)
    MDL Number MFCD00149725
    CAS 13780-42-2
    Melting Point 105° to 110°C
    TSCA TSCA
    Odor Odorless
    Physical Form Crystalline Aggregates
    Sensitivity Hygroscopic
    Quantity 10g
    Density 3.860g/mL
    Chemical Name or Material Gallium(III) sulfate hydrate
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit