Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallium(III) sulfide, 99.99% (metals basis)

Catalog No. p-7049220
Click to view available options
Quantity:
0.5 g
2 g
10 g

CAS: 12024-22-5 | Ga2S3 | 235.63 g/mol

Gallium(III) sulfide is used in electronics and photonics. It acts as an active material in lithium secondary batteries. Further, it is used in thin-film solar cells. It is also used to prepare gallium lanthanum sulfide glass by reacting with lanthanum sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12024-22-5
Molecular Formula Ga2S3
Molecular Weight (g/mol) 235.63
MDL Number MFCD00016106
InChI Key BVSHTEBQPBBCFT-UHFFFAOYSA-N
IUPAC Name digallium(3+) trisulfanediide
SMILES [S--].[S--].[S--].[Ga+3].[Ga+3]

Specifications

Melting Point 1255°C
Physical Form Powder
Quantity 0.5 g
UN Number UN3260
Formula Weight 235.63
Percent Purity 99.99%
Odor Odorless
Sensitivity Moisture sensitive
Density 3.77 g/mL
Assay (metals basis)
Chemical Name or Material Gallium(III) sulfide
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 234-692-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.