Learn More
Gallium Indium alloy, 99.99% (metals basis)
Description
Step soldering involving temperature sensitive components, Soldering to molded interconnect device (MID) plastics, Fusible alloys/fuse applications, Mercury replacement, Thermal and electrical conductivity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Gray |
| Chemical Name or Material | Gallium Indium alloy |
| Quantity | 50 g |
| Odor | Odorless |
| Form | Solid |
| Assay Percent Range | 99.99% (metals basis) |
| Molecular Formula | Ga:In; 40:60 wt.% |
| MDL Number | MFCD00144387 |
| Solubility Information | Insoluble in water. |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.