Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallium Indium eutectic, 99.99% (metals basis)

Catalog No. p-4408795
Click to view available options
Quantity:
1 g
10 g
50 g

Gallium Indium eutectic, Ga:In; 75.5:24.5 wt%, 10 g, 99.99% (metals basis), 6.2, Hazard Class: 8; Packaging Group: III, 231-163-8, H290-H314-H335, P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c, 15.7 deg.C, MFCD00144387, Liquid | Ga:In; 75.5:24.5 wt%

It useful as a contact electrode for the electrical characterization of thin film organic and semiconductor devices. Gallium-Indium eutectic top contact, and a halogen light source was used to test P3HT-PCBM organic solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gallium Indium eutectic
Melting Point 15.7°C
Density 6.2
Quantity 1 g
Form Liquid
Assay Percent Range 99.99% (metals basis)
Molecular Formula Ga:In; 75.5:24.5 wt%
MDL Number MFCD00144387
Solubility Information Insoluble in water or organic solvents.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.