Gallium Indium eutectic, 99.99% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1247803

Quantity 1g

 View more versions of this product

Catalog No. AA1247803

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Liquid
    Assay Percent Range 99.99% (metals basis)
    Molecular Formula Ga:In; 75.5:24.5 wt%
    MDL Number MFCD00144387
    Chemical Name or Material Gallium Indium eutectic

    Specifications

    Quantity 1g
    EINECS Number 231-163-8
    Physical Form Liquid
    Assay Percent Range 99.99% (metals basis)
    Molecular Formula Ga:In; 75.5:24.5 wt%
    MDL Number MFCD00144387
    Chemical Name or Material Gallium Indium eutectic

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit