Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallium metal, packaged in polyethylene bottle, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number AA4584136
Click to view available options
Quantity:
100 g
500 g
7440-55-3
Ga
69.72
MFCD00134045
GYHNNYVSQQEPJS-UHFFFAOYSA-N
gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom
5360835
CHEBI:49631
gallium
[Ga]
7440-55-3
Ga
69.72
MFCD00134045
GYHNNYVSQQEPJS-UHFFFAOYSA-N
gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom
5360835
CHEBI:49631
gallium
[Ga]

May be solid or liquid due to ambient temperature

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver to Gray
Appearance Silver Gray
Assay Percent Range 99.9999% (Metals basis)
Quantity 500 g
Form Solid and/or Liquid
Chemical Name or Material Gallium
Hazard Category H290-H314-H335
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 231-163-8
RTECSNumber LW8600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit