Gallium metal, 99.999% (metals basis), Alfa Aesar™

May be solid or liquid due to ambient temperature

Manufacturer: Alfa Aesar 4441836

Quantity 500g

 View more versions of this product

Catalog No. AA4441836

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-55-3
    Physical Form Metal
    Assay Percent Range 99.9999% (Metals basis)
    MDL Number MFCD00134045
    Chemical Name or Material Gallium

    Specifications

    Quantity 500g
    Color Silver-gray
    DOT Information DOT Class: 8, Packaging Group: III
    EINECS Number 231-163-8
    Physical Form Metal
    Assay Percent Range 99.9999% (Metals basis)
    MDL Number MFCD00134045
    CAS 7440-55-3
    Appearance Silver Gray
    Chemical Name or Material Gallium

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit