Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gallium Tin alloy, 99.99% (metals basis)
Click to view available options
Quantity:
1 g
10 g
50 g
Description
Gallium Tin alloy is used to cool computer chips in place of water. Used in medical thermometers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Silver |
Melting Point | ≥25°C |
Appearance | Silver-colored liquid |
Quantity | 10 g |
Form | Melting Solid |
Assay Percent Range | 99.99% (metals basis) |
Solubility Information | Insoluble in water. |
Concentration or Composition (by Analyte or Components) | Tin: 8.0%; Gallium: 92.0% |
Chemical Name or Material | Gallium Tin alloy |
Safety and Handling
Hazard Category | H290-H314-H335 |
Hazard Statement | Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. |
Precautionary Statement | P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c |
DOTInformation | Hazard Class: 8, Packing Group: III |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction