Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GALM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557285
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557285 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557285 Supplier Invitrogen™ Supplier No. PA557285
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PVELGKHLQD FHLNGFDHNF CLKGSKEKHF CARVHHAASG RVLEVYTTQP GVQFYTGNFL DGTLKGKNGA VYPKHSGFCL ETQNWPDAVN QPRFP Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.

This gene encodes an enzyme that catalyzes the epimerization of hexose sugars such as glucose and galactose. The encoded protein is expressed in the cytoplasm and has a preference for galactose. The encoded protein may be required for normal galactose metabolism by maintaining the equilibrium of alpha and beta anomers of galactose.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALM
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GALM
Gene Accession No. Q96C23
Gene Alias A530057M15Rik; aldose 1-epimerase; aldose mutarotase; AU015645; AU020959; BLOCK25; epididymis secretory sperm binding protein Li 63p; galactomutarotase; galactose enzyme activator; galactose mutarotase; galactose mutarotase (aldose 1-epimerase); GALM; GLAT; HEL-S-63p; IBD1
Gene Symbols GALM
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GALM. Recombinant protein control fragment (Product #RP-102665).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130589
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.