Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GALNS Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518808
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
This gene encodes N-acetylgalactosamine-6-sulfatase whichis a lysosomal exohydrolase required for the degradation of theglycosaminoglycans, keratan sulfate, and chondroitin 6-sulfate.Sequence alterations including point, missense and nonsensemutations, as well as those that affect splicing, result in adeficiency of this enzyme. Deficiencies of this enzyme lead toMorquio A syndrome, a lysosomal storage disorder.
Specifications
GALNS | |
Polyclonal | |
Unconjugated | |
GALNS | |
Chondroitinase; chondroitinsulfatase; galactosamine (N-acetyl)-6-sulfatase; galactosamine (N-acetyl)-6-sulfate sulfatase; galactose-6-sulfate sulfatase; GalN6S; GALNAC6S; GalNAc6S sulfatase; Galns; GAS; mFLJ00319; MPS4A; N-acetylgalactosamine-6-sulfatase; N-acetylgalactosamine-6-sulfate sulfatase | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
2588 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P34059 | |
GALNS | |
Synthetic peptide sequence (TTHNLEDHTKLP) corresponding to the internal amino acids of GALNS (aa 428-440). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction