Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GALNS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115267
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115267 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115267 Supplier Invitrogen™ Supplier No. PA5115267
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GALNS.

This gene encodes N-acetylgalactosamine-6-sulfatase whichis a lysosomal exohydrolase required for the degradation of theglycosaminoglycans, keratan sulfate, and chondroitin 6-sulfate.Sequence alterations including point, missense and nonsensemutations, as well as those that affect splicing, result in adeficiency of this enzyme. Deficiencies of this enzyme lead toMorquio A syndrome, a lysosomal storage disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALNS
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene GALNS
Gene Accession No. P34059, Q32KJ6, Q571E4
Gene Alias Chondroitinase; chondroitinsulfatase; galactosamine (N-acetyl)-6-sulfatase; galactosamine (N-acetyl)-6-sulfate sulfatase; galactose-6-sulfate sulfatase; GalN6S; GALNAC6S; GalNAc6S sulfatase; Galns; GAS; mFLJ00319; MPS4A; N-acetylgalactosamine-6-sulfatase; N-acetylgalactosamine-6-sulfate sulfatase
Gene Symbols GALNS
Host Species Rabbit
Immunogen A synthesized peptide derived from human GALNS(Accession P34059), corresponding to amino acid residues S408-S458.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2588, 292073, 50917
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.