Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GALNT10/GalNAc-T10 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982757
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2757 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2757 Supplier Bioss Supplier No. BS13270R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Protein glycosylation is an important biological process that is carried out by a large family of glycosyltransferases that catalyze the synthesis of oligosaccharides and glycoconjugates. Polypeptide GalNAc transferases initiate the synthesis of mucin-type oligosaccharides by transferring GalNAc from UDP-GalNAc to the hydroxyl group of either a serine or threonine residue on the polypeptide acceptor. Polypeptide galactoaminyltransferase 10 (GALNT10) belongs to the polypeptide N-acetylgalactosaminyl-transferase (pp-GalNAc-T) protein family. Following expression in insect cells, recombinant GALNT10 showed significant GalNAcT activity toward mucin-derived peptides, and it utilized both non-glycosylated and glycosylated peptide substrates. GALNT10 mRNA is highly expressed in several distinct hypothalamic, thalamic, and amygdaloid nuclei in mouse brain. At least four isoforms of GALNT10 are known to exist.

Specifications

Antigen GALNT10/GalNAc-T10
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GALNT10
Gene Accession No. Q86SR1
Gene Alias AU018154; C330012K04Rik; GalNAc transferase 10; GALNACT10; GalNAc-T10; GalNAc-T9; GALNT10; Galnt9; Polypeptide GalNAc transferase 10; Polypeptide N-acetylgalactosaminyltransferase 10; PP PPGANTASE10; PPGALNACT10; ppGaNTase; pp-GaNTase 10; PPGANTASE10; Protein-UDP acetylgalactosaminyltransferase 10; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 10; UDP-N-acetyl-alpha-D-galactosamine: polypeptide N-acetylgalactosaminyltransferase 9; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 10; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 10 (GalNAc-T10); UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 9
Gene Symbols GALNT10
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GALNT10/GalNAc-T10.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55568
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.