Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GALNT11/GalNAc-T11 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS13271R

Description
Polypeptide N-acetylgalactosaminyltransferase that catalyzes the initiation of protein O-linked glycosylation and is involved in left/right asymmetry by mediating O-glycosylation of NOTCH1. O-glycosylation of NOTCH1 promotes activation of NOTCH1, modulating the balance between motile and immotile (sensory) cilia at the left-right organiser (LRO). Polypeptide N- acetylgalactosaminyltransferases catalyze the transfer of an N- acetyl-D-galactosamine residue to a serine or threonine residue on the protein receptor. Displays the same enzyme activity toward MUC1, MUC4, and EA2 than GALNT1. Not involved in glycosylation of erythropoietin (EPO).Specifications
GALNT11/GalNAc-T11 | |
Polyclonal | |
Unconjugated | |
GALNT11 | |
GALNACT11; GalNAc-T11; GALNT11; Polypeptide GalNAc transferase 11; Polypeptide N-acetylgalactosaminyltransferase 11; pp-GaNTase 11; Protein-UDP acetylgalactosaminyltransferase 11; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 11; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 11 (GalNAc-T11) | |
Rabbit | |
Protein A | |
RUO | |
63917 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q8NCW6 | |
GALNT11 | |
KLH conjugated synthetic peptide derived from human GALNT11/GalNAc-T11. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction