Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GALNT11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558738
Description
Immunogen sequence: LKTKSYGNIS ERVELRKKLG CKSFKWYLDN VYPEMQISGS HAKPQQPIFV NRGPKRPKVL QRGRLYHLQT NKCLVAQGRP SQKGGLVVLK ACDYSDPNQI WIYNEEHELV LNSLLCLDMS ETRSSDPPRL MKCHGSG Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 87%.
Polypeptide N-acetylgalactosaminyltransferase that catalyzes the initiation of protein O-linked glycosylation and is involved in left/right asymmetry by mediating O-glycosylation of NOTCH1. O-glycosylation of NOTCH1 promotes activation of NOTCH1, modulating the balance between motile and immotile (sensory) cilia at the left-right organiser (LRO). Polypeptide N- acetylgalactosaminyltransferases catalyze the transfer of an N- acetyl-D-galactosamine residue to a serine or threonine residue on the protein receptor. Displays the same enzyme activity toward MUC1, MUC4, and EA2 than GALNT1. Not involved in glycosylation of erythropoietin (EPO).
Specifications
GALNT11 | |
Polyclonal | |
Unconjugated | |
GALNT11 | |
:polypeptide N-acetylgalactosaminyltransferase 11; A430075I06Rik; AI648252; E430002F06Rik; GALNACT11; GALNAC-T11; Galnt11; Polypeptide GalNAc transferase 11; polypeptide N-acetylgalactosaminyltransferase 11; pp-GaNTase 11; protein-UDP acetylgalactosaminyltransferase 11; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 11; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 11; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 11 (GalNAc-T11) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
63917 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NCW6 | |
GALNT11 | |
Recombinant protein corresponding to Human GALNT11. Recombinant protein control fragment (Product #RP-90498). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction