Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GALNT12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561701
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561701 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561701 Supplier Invitrogen™ Supplier No. PA561701
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEGCIAVEAG MDTLIMHLCE ETAPENQKFI LQEDGSLFHE QSKKCVQAAR KESSDSFVPL LRDCTNSDHQ K Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 59%.

GALNT12 is a member of a family of UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferases (EC 2.4.1.41), which catalyze the transfer of N-acetylgalactosamine (GalNAc) from UDP-GalNAc to a hydroxyl amino acid on a polypeptide acceptor in the initial step of mucin-type O-linked protein glycosylation (Guo et al., 2002 [PubMed 12135769]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALNT12
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GALNT12
Gene Accession No. Q8IXK2
Gene Alias colorectal cancer, susceptibility to 1; CRCS1; GalNAc-T12; GALNT12; Polypeptide GalNAc transferase 12; polypeptide N-acetylgalactosaminyltransferase 12; pp-GaNTase 12; protein-UDP acetylgalactosaminyltransferase 12; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 12; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 12 (GalNAc-T12)
Gene Symbols GALNT12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GALNT12. Recombinant protein control fragment (Product #RP-100931).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79695
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.