Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GALNT12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144997
Description
Immunogen sequence: VPEDRPGFFG MLQNKGLTDY CFDYNPPDEN QIVGHQVILY LCHGMGQNQF F Antigen sequence identity: Mouse 82%; Rat 80%.
GALNT12 is a member of a family of UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferases (EC 2.4.1.41), which catalyze the transfer of N-acetylgalactosamine (GalNAc) from UDP-GalNAc to a hydroxyl amino acid on a polypeptide acceptor in the initial step of mucin-type O-linked protein glycosylation (Guo et al., 2002 [PubMed 12135769]).
Specifications
GALNT12 | |
Polyclonal | |
Unconjugated | |
GALNT12 | |
colorectal cancer, susceptibility to 1; CRCS1; GalNAc-T12; GALNT12; Polypeptide GalNAc transferase 12; polypeptide N-acetylgalactosaminyltransferase 12; pp-GaNTase 12; protein-UDP acetylgalactosaminyltransferase 12; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 12; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 12 (GalNAc-T12) | |
Rabbit | |
Affinity chromatography | |
RUO | |
79695 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IXK2 | |
GALNT12 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 438-488. Recombinant protein control fragment (Product #RP-110038). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction