Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GALNT12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531582
Description
Recommended positive controls: A549. Predicted reactivity: Mouse (89%), Rhesus Monkey (96%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GALNT12 is a member of a family of UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferases (EC 2.4.1.41), which catalyze the transfer of N-acetylgalactosamine (GalNAc) from UDP-GalNAc to a hydroxyl amino acid on a polypeptide acceptor in the initial step of mucin-type O-linked protein glycosylation (Guo et al., 2002 [PubMed 12135769]).
Specifications
| GALNT12 | |
| Polyclonal | |
| Unconjugated | |
| GALNT12 | |
| colorectal cancer, susceptibility to 1; CRCS1; GalNAc-T12; GALNT12; Polypeptide GalNAc transferase 12; polypeptide N-acetylgalactosaminyltransferase 12; pp-GaNTase 12; protein-UDP acetylgalactosaminyltransferase 12; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 12; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 12 (GalNAc-T12) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79695 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.63 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8IXK2 | |
| GALNT12 | |
| Recombinant protein encompassing a sequence within the center region of human GALNT12. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction