Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GALNT13/GalNAc-T13 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982760
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2760 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2760 Supplier Bioss Supplier No. BS13274R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The GALNT13 protein is a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAcT; EC 2.4.1.41) family, which initiate O-linked glycosylation of mucins by the initial transfer of N-acetylgalactosamine (GalNAc) with an alpha-linkage to a serine or threonine residue.The GALNT13 protein is a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAcT; EC 2.4.1.41) family, which initiate O-linked glycosylation of mucins (see MUC3A, MIM 158371) by the initial transfer of N-acetylgalactosamine (GalNAc) with an alpha-linkage to a serine or threonine residue.[supplied by OMIM].

Specifications

Antigen GALNT13/GalNAc-T13
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GALNT13
Gene Accession No. Q8IUC8
Gene Alias A230002A12; A230020F20; BB182356; BOS_1764; FLJ16031; FLJ41157; GalNAc transferase 13; GalNAc-T13; GALNT13; H_NH0187G20.1; KIAA1918; LOC100353091; MGC119459; MGC119461; Polypeptide GalNAc transferase 13; Polypeptide N-acetylgalactosaminyltransferase 13; polypeptide N-acetylgalactosaminyltransferase 13-like; pp-GalNAc-T13; pp-GaNTase 13; Protein-UDP acetylgalactosaminyltransferase 13; T13; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 13; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 13; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 13 (GalNAc-T13); WUGSC:H_NH0187G20.1
Gene Symbols GALNT13
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GALNT13/GalNAc-T13.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114805
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.